Product Details

SNP ID
rs115467558
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:119621276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAATGCAAAAGGTCAGGCTACA[A/G]TTATTCACACTGTCCTCAGCCTCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF697 PubMed Links

Gene Details

Gene
ZNF697
Gene Name
zinc finger protein 697
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080470.1 6186 UTR 3 NP_001073939.1
XM_005271315.3 6186 UTR 3 XP_005271372.1
XM_011542416.2 6186 UTR 3 XP_011540718.1
XM_017002783.1 6186 UTR 3 XP_016858272.1

View Full Product Details