Product Details

SNP ID
rs115493951
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:241534415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATAACATGAGATTGTCATCATG[C/T]AACCACGTCATACATATGCTGGAAA
Phenotype
MIM: 603538
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KMO PubMed Links

Gene Details

Gene
KMO
Gene Name
kynurenine 3-monooxygenase (kynurenine 3-hydroxylase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003679.4 Intron NP_003670.2
XM_005273337.1 Intron XP_005273394.1
XM_005273338.3 Intron XP_005273395.1
XM_017002649.1 Intron XP_016858138.1
XM_017002650.1 Intron XP_016858139.1

View Full Product Details