Product Details

SNP ID
rs115502426
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:183260028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTTTGGGTTTGTGGTAGAAACT[G/T]GTAACTGGCCCCTAATACCCATTCT
Phenotype
MIM: 608701
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NMNAT2 PubMed Links

Gene Details

Gene
NMNAT2
Gene Name
nicotinamide nucleotide adenylyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015039.3 Intron NP_055854.1
NM_170706.3 Intron NP_733820.1

View Full Product Details