Product Details

SNP ID
rs115541153
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:201468609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGTGCTCTCCACGCACTCCACGG[A/C]CTTGATGCGGGCGAAGCTGAGCTCC
Phenotype
MIM: 607054
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PHLDA3 PubMed Links

Gene Details

Gene
PHLDA3
Gene Name
pleckstrin homology like domain family A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012396.4 563 Missense Mutation GCC,TCC A60S NP_036528.1

View Full Product Details