Product Details

SNP ID
rs115545469
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:47183786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGGGCCACAGCCGAGCCCCAA[C/T]CTAGACACGGTCTGAGCTCCAACCT
Phenotype
MIM: 607178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00853 PubMed Links

Gene Details

Gene
LINC00853
Gene Name
long intergenic non-protein coding RNA 853
There are no transcripts associated with this gene.

Gene
PDZK1IP1
Gene Name
PDZK1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005764.3 697 UTR 3 NP_005755.1

View Full Product Details