Product Details

SNP ID
rs115598876
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:23629102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACTCAGAGAAAATCTTAACACC[G/T]TTCTGTGGCCTAGAAAGTCCTACCT
Phenotype
MIM: 607305
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MDS2 PubMed Links

Gene Details

Gene
MDS2
Gene Name
myelodysplastic syndrome 2 translocation associated
There are no transcripts associated with this gene.

View Full Product Details