Product Details

SNP ID
rs115611466
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:2393685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAGCCTGTCCTAGTCACACTGG[C/T]AGAGAGTGGTAGAGCCAGGATCTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MORN1 PubMed Links

Gene Details

Gene
MORN1
Gene Name
MORN repeat containing 1
There are no transcripts associated with this gene.

Gene
RER1
Gene Name
retention in endoplasmic reticulum sorting receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007033.4 Intron NP_008964.3
XM_005244713.4 Intron XP_005244770.2
XM_006710306.2 Intron XP_006710369.1
XM_011540542.1 Intron XP_011538844.1
XM_011540543.1 Intron XP_011538845.1
XM_017000131.1 Intron XP_016855620.1
XM_017000132.1 Intron XP_016855621.1
XM_017000133.1 Intron XP_016855622.1
XM_017000134.1 Intron XP_016855623.1
XM_017000135.1 Intron XP_016855624.1

View Full Product Details