Product Details
- SNP ID
-
rs115633843
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:153150087 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGAGGTGGCTGGCAGGGCTGCTT[A/G]CACTGCTGCTGCTGGTAAGACATCT
- Phenotype
-
MIM: 182269
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SPRR2C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs513555] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPRR2C
- Gene Name
- small proline rich protein 2C (pseudogene)
There are no transcripts associated with this gene.
- Gene
- SPRR2G
- Gene Name
- small proline rich protein 2G
View Full Product Details