Product Details

SNP ID
rs115638937
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:152827858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGGTGAAATGAGTGATCAAACA[A/T]TTCCTCCTCACCTGTCTCCTCCTGG
Phenotype
MIM: 612603
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LCE1A PubMed Links

Gene Details

Gene
LCE1A
Gene Name
late cornified envelope 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178348.2 386 UTR 3 NP_848125.1

View Full Product Details