Product Details

SNP ID
rs115757443
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:185062881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACTCATGATTGAGGCTTGCCAA[G/T]TAAAACAACAAAGAAGTATCATTTC
Phenotype
MIM: 608985
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RNF2 PubMed Links

Gene Details

Gene
RNF2
Gene Name
ring finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007212.3 Intron NP_009143.1
XM_005245413.3 Intron XP_005245470.1
XM_011509851.2 Intron XP_011508153.1
XM_011509852.2 Intron XP_011508154.1

View Full Product Details