Product Details

SNP ID
rs115790260
Assay Type
Functionally Tested
NCBI dbSNP Submissions
16
Location
Chr.1:225815930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTTAATGGTGGGAAGGAACTCC[A/G]GTAAGTGTGTTATATGATCTGCTCC
Phenotype
MIM: 132810
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
EPHX1 PubMed Links

Gene Details

Gene
EPHX1
Gene Name
epoxide hydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000120.3 Intron NP_000111.1
NM_001136018.3 Intron NP_001129490.1
NM_001291163.1 Intron NP_001278092.1

View Full Product Details