Product Details

SNP ID
rs115820761
Assay Type
Functionally Tested
NCBI dbSNP Submissions
8
Location
Chr.1:156054485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGAGAAAACTGTTTTGAAAGTAA[C/T]CAAAGGCGTGCCTGACAAATATGAG
Phenotype
MIM: 610389 MIM: 612942 MIM: 605440
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LAMTOR2 PubMed Links

Gene Details

Gene
LAMTOR2
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145264.1 Intron NP_001138736.1
NM_014017.3 Intron NP_054736.1
Gene
RAB25
Gene Name
RAB25, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
UBQLN4
Gene Name
ubiquilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304342.1 Intron NP_001291271.1
NM_020131.4 Intron NP_064516.2
XM_005245348.3 Intron XP_005245405.1

View Full Product Details