Product Details

SNP ID
rs115824339
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:25246498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGCGACCTCGTCCTGGATCTGT[C/T]CCTCCATCTGCTGTGCTCCTCCGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSRP1 PubMed Links

Gene Details

Gene
RSRP1
Gene Name
arginine and serine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321772.1 708 Missense Mutation AAC,GAC N156D NP_001308701.1
NM_020317.4 708 Missense Mutation AAC,GAC N156D NP_064713.3
XM_011541797.1 708 Missense Mutation AAC,GAC N156D XP_011540099.1

View Full Product Details