Product Details
- SNP ID
-
rs115825348
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:20664650 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTGGCCTTGGTGAAAGGGATGTC[A/G]AGGGACTCGAGGCGGAAGGGCCGGG
- Phenotype
-
MIM: 602202
MIM: 605037
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DDOST
PubMed Links
Gene Details
- Gene
- DDOST
- Gene Name
- dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit
There are no transcripts associated with this gene.
- Gene
- KIF17
- Gene Name
- kinesin family member 17
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001122819.2 |
3332 |
Silent Mutation |
CTC,CTT |
L1007L |
NP_001116291.1 |
NM_001287212.1 |
3332 |
Silent Mutation |
CTC,CTT |
L908L |
NP_001274141.1 |
NM_020816.3 |
3332 |
Silent Mutation |
CTC,CTT |
L1008L |
NP_065867.2 |
XM_005245950.3 |
3332 |
Intron |
|
|
XP_005246007.1 |
XM_005245951.3 |
3332 |
Intron |
|
|
XP_005246008.1 |
XM_011541837.1 |
3332 |
Missense Mutation |
TCG,TTG |
S1007L |
XP_011540139.1 |
XM_011541838.1 |
3332 |
Missense Mutation |
TCG,TTG |
S1006L |
XP_011540140.1 |
XM_011541839.2 |
3332 |
Silent Mutation |
CTC,CTT |
L1008L |
XP_011540141.1 |
XM_011541840.2 |
3332 |
Silent Mutation |
CTC,CTT |
L1007L |
XP_011540142.1 |
XM_011541841.2 |
3332 |
Missense Mutation |
TCG,TTG |
S1007L |
XP_011540143.1 |
XM_011541842.1 |
3332 |
Silent Mutation |
CTC,CTT |
L1008L |
XP_011540144.1 |
XM_011541843.1 |
3332 |
Intron |
|
|
XP_011540145.1 |
XM_011541844.2 |
3332 |
Intron |
|
|
XP_011540146.1 |
XM_011541845.2 |
3332 |
Intron |
|
|
XP_011540147.1 |
XM_011541846.2 |
3332 |
Missense Mutation |
TCG,TTG |
S907L |
XP_011540148.1 |
XM_011541847.2 |
3332 |
Missense Mutation |
TCG,TTG |
S533L |
XP_011540149.1 |
View Full Product Details