Product Details

SNP ID
rs115829992
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:159001597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACATGAACGCCCCTGTCTTACC[A/C]GTGAAAGAAAAGAAGGAAATTTGCA
Phenotype
MIM: 147586 MIM: 612677
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IFI16 PubMed Links

Gene Details

Gene
IFI16
Gene Name
interferon gamma inducible protein 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206567.1 Intron NP_001193496.1
NM_005531.2 Intron NP_005522.2
XM_005245127.3 Intron XP_005245184.1
XM_006711290.1 Intron XP_006711353.1
XM_017001149.1 Intron XP_016856638.1
XM_017001150.1 Intron XP_016856639.1
Gene
POP3
Gene Name
pyrin domain-only protein 3
There are no transcripts associated with this gene.

Gene
PYHIN1
Gene Name
pyrin and HIN domain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152501.4 Intron NP_689714.2
NM_198928.4 Intron NP_945146.1
NM_198929.4 Intron NP_945147.1
NM_198930.3 Intron NP_945148.1
XM_005244930.1 Intron XP_005244987.1
XM_011509242.2 Intron XP_011507544.1
XM_011509243.2 Intron XP_011507545.1
XM_017000463.1 Intron XP_016855952.1

View Full Product Details