Product Details

SNP ID
rs115834094
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:42154821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCTCTGCCTCAGGACCTTCAG[C/T]CTGTCTGCGCCTCGCAGGAGGCTTC
Phenotype
MIM: 139392 MIM: 601271
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCA2A PubMed Links

Gene Details

Gene
GUCA2A
Gene Name
guanylate cyclase activator 2A
There are no transcripts associated with this gene.

Gene
GUCA2B
Gene Name
guanylate cyclase activator 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007102.2 262 Missense Mutation CCT,TCT P78S NP_009033.1

View Full Product Details