Product Details

SNP ID
rs115838697
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:1174787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGTCCCACCTGCATTCAGGCCCC[A/G]GGTTCACCACACACACAGCGGGCAG
Phenotype
MIM: 612090 MIM: 612091 MIM: 612094
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR200A PubMed Links

Gene Details

Gene
MIR200A
Gene Name
microRNA 200a
There are no transcripts associated with this gene.

Gene
MIR200B
Gene Name
microRNA 200b
There are no transcripts associated with this gene.

Gene
MIR429
Gene Name
microRNA 429
There are no transcripts associated with this gene.

Gene
TTLL10
Gene Name
tubulin tyrosine ligase like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130045.1 Intron NP_001123517.1
NM_153254.2 Intron NP_694986.2
XM_005244738.1 Intron XP_005244795.1
XM_011541177.2 Intron XP_011539479.1
XM_017000906.1 Intron XP_016856395.1
XM_017000907.1 Intron XP_016856396.1
XM_017000908.1 Intron XP_016856397.1
XM_017000909.1 Intron XP_016856398.1
XM_017000910.1 Intron XP_016856399.1
XM_017000911.1 Intron XP_016856400.1
XM_017000912.1 Intron XP_016856401.1
Gene
TTLL10-AS1
Gene Name
TTLL10 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details