Product Details

SNP ID
rs115906127
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:53248945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTACAAATAACAGTGAGCAGAA[A/G]GAAGTGACTGAGATCACAGGGCAGT
Phenotype
MIM: 602600
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100507564 PubMed Links

Gene Details

Gene
LOC100507564
Gene Name
uncharacterized LOC100507564
There are no transcripts associated with this gene.

Gene
LRP8
Gene Name
LDL receptor related protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018054.2 Intron NP_001018064.1
NM_004631.4 Intron NP_004622.2
NM_017522.4 Intron NP_059992.3
NM_033300.3 Intron NP_150643.2
XM_005271173.3 Intron XP_005271230.1
XM_005271174.3 Intron XP_005271231.1
XM_005271175.3 Intron XP_005271232.1
XM_006710881.3 Intron XP_006710944.1
XM_006710882.3 Intron XP_006710945.1
XM_011542094.2 Intron XP_011540396.1
XM_011542095.2 Intron XP_011540397.1
XM_011542096.2 Intron XP_011540398.1
XM_017002265.1 Intron XP_016857754.1
XM_017002266.1 Intron XP_016857755.1
XM_017002267.1 Intron XP_016857756.1
XM_017002268.1 Intron XP_016857757.1

View Full Product Details