Product Details

SNP ID
rs115972666
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:43384469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCTTCACCAGAGCTGCAGGTGG[A/G]CATTTTTTTTTCCTTCCTGGCCCAG
Phenotype
MIM: 607956 MIM: 615463
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED8 PubMed Links

Gene Details

Gene
MED8
Gene Name
mediator complex subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001653.2 935 UTR 3 NP_001001653.1
NM_052877.4 935 Missense Mutation GCC,GTC A296V NP_443109.2
NM_201542.4 935 UTR 3 NP_963836.2
Gene
SZT2
Gene Name
seizure threshold 2 homolog (mouse)
There are no transcripts associated with this gene.

View Full Product Details