Product Details

SNP ID
rs116119975
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:45328052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAGGCAGATCCGAGCTGCAGGG[C/T]ACGAGCCTCATCTGCTTGCTCGACA
Phenotype
MIM: 604933
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HPDL PubMed Links

Gene Details

Gene
HPDL
Gene Name
4-hydroxyphenylpyruvate dioxygenase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032756.2 1180 Missense Mutation CAC,TAC H302Y NP_116145.1
Gene
MUTYH
Gene Name
mutY DNA glycosylase
There are no transcripts associated with this gene.

View Full Product Details