Product Details

SNP ID
rs116120838
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:151800417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCCATCCTCTTCTCTGTAGCCT[A/G]TGGGTCCTTTGCTCTTATTCCTGTA
Phenotype
MIM: 609794 MIM: 602943 MIM: 609501
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINGO4 PubMed Links

Gene Details

Gene
LINGO4
Gene Name
leucine rich repeat and Ig domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004432.3 2514 UTR 3 NP_001004432.1
Gene
RORC
Gene Name
RAR related orphan receptor C
There are no transcripts associated with this gene.

Gene
TDRKH
Gene Name
tudor and KH domain containing
There are no transcripts associated with this gene.

View Full Product Details