Product Details

SNP ID
rs116158450
Assay Type
Functionally Tested
NCBI dbSNP Submissions
17
Location
Chr.1:23748048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTAGAGAAAGGTTTGGCCAGTC[A/C]TAACTTTTGTTAGAGCAAGTAGGAA
Phenotype
MIM: 600786
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
TCEB3 PubMed Links

Gene Details

Gene
TCEB3
Gene Name
transcription elongation factor B subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003198.2 Intron NP_003189.2

View Full Product Details