Product Details

SNP ID
rs116242087
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:26476638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACAGGAATAAAGTAAATTGGAG[A/G]CTAATTGCTTTGTCCATTCCCTTAG
Phenotype
MIM: 608172 MIM: 163910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHDDS PubMed Links

Gene Details

Gene
DHDDS
Gene Name
dehydrodolichyl diphosphate synthase subunit
There are no transcripts associated with this gene.

Gene
HMGN2
Gene Name
high mobility group nucleosomal binding domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005517.3 1965 UTR 3 NP_005508.1
Gene
LOC101928324
Gene Name
uncharacterized LOC101928324
There are no transcripts associated with this gene.

View Full Product Details