Product Details

SNP ID
rs116327280
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:204423610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGAAAAATCATGGGGCAAAAA[A/G]CAACAGTTTTGATGGAATCCCCCAC
Phenotype
MIM: 602838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIK3C2B PubMed Links

Gene Details

Gene
PIK3C2B
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002646.3 5572 UTR 3 NP_002637.3
XM_005245257.2 5572 UTR 3 XP_005245314.1
XM_005245258.3 5572 UTR 3 XP_005245315.1
XM_011509630.2 5572 UTR 3 XP_011507932.1
XM_011509631.2 5572 UTR 3 XP_011507933.1
XM_011509633.2 5572 Intron XP_011507935.1
XM_017001473.1 5572 UTR 3 XP_016856962.1
XM_017001474.1 5572 UTR 3 XP_016856963.1
XM_017001475.1 5572 UTR 3 XP_016856964.1

View Full Product Details