Product Details

SNP ID
rs116356405
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:116576989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGTACATTACAAAGAATAAGAA[G/T]CCCTGTAACATCTATCTGAGAATAC
Phenotype
MIM: 153420 MIM: 603491
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD58 PubMed Links

Gene Details

Gene
CD58
Gene Name
CD58 molecule
There are no transcripts associated with this gene.

Gene
IGSF3
Gene Name
immunoglobulin superfamily member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007237.2 4598 UTR 3 NP_001007238.1
NM_001542.3 4598 UTR 3 NP_001533.2
XM_005270793.2 4598 UTR 3 XP_005270850.1
XM_005270794.4 4598 UTR 3 XP_005270851.1
XM_006710593.3 4598 UTR 3 XP_006710656.1
XM_011541315.1 4598 UTR 3 XP_011539617.1
XM_011541316.2 4598 UTR 3 XP_011539618.1

View Full Product Details