Product Details

SNP ID
rs116394482
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:23361527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTGTGTGCCCAACTACTATGGT[A/G]TGTGAATAGGTGTGGCAATTTAGGC
Phenotype
MIM: 611703
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF436 PubMed Links

Gene Details

Gene
ZNF436
Gene Name
zinc finger protein 436
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077195.1 2414 UTR 3 NP_001070663.1
NM_030634.2 2414 UTR 3 NP_085137.1
XM_011542201.1 2414 UTR 3 XP_011540503.1
Gene
ZNF436-AS1
Gene Name
ZNF436 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details