Product Details

SNP ID
rs116473357
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:169132726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATAAAAGAATGAACACATTCCTC[A/G]TGTGTGATTCACTCTTGTCTAAATG
Phenotype
MIM: 182330 MIM: 613465
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP1B1 PubMed Links

Gene Details

Gene
ATP1B1
Gene Name
ATPase Na+/K+ transporting subunit beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001677.3 1009 Intron NP_001668.1
Gene
NME7
Gene Name
NME/NM23 family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013330.4 1009 UTR 3 NP_037462.1
NM_197972.2 1009 UTR 3 NP_932076.1
XM_005245106.4 1009 UTR 3 XP_005245163.1
XM_017001099.1 1009 Intron XP_016856588.1
XM_017001100.1 1009 UTR 3 XP_016856589.1

View Full Product Details