Product Details

SNP ID
rs116632678
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:185741895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGCCGTGTCCAACTACTGGAACA[A/G]TTGCTTTGTTCATGAGGCTAAAACT
Phenotype
MIM: 608548
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMCN1 PubMed Links

Gene Details

Gene
HMCN1
Gene Name
hemicentin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031935.2 Intron NP_114141.2
XM_011510038.2 Intron XP_011508340.1
XM_011510041.2 Intron XP_011508343.1
XM_017002437.1 Intron XP_016857926.1

View Full Product Details