Product Details

SNP ID
rs116719577
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:26411441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGGAGGCGCCGGAGGACGCGGC[C/G]CGGGCGGCGGACGAGCCTCAGCTGC
Phenotype
MIM: 611043
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LIN28A PubMed Links

Gene Details

Gene
LIN28A
Gene Name
lin-28 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024674.5 Intron NP_078950.1
XM_011542148.1 Intron XP_011540450.1

View Full Product Details