Product Details

SNP ID
rs116785219
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:32028940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCATTGCCACCTCTATAGTCTGG[G/T]AACTCCAGTATATAAACAGTGAAGA
Phenotype
MIM: 602489
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KHDRBS1 PubMed Links

Gene Details

Gene
KHDRBS1
Gene Name
KH RNA binding domain containing, signal transduction associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271878.1 Intron NP_001258807.1
NM_006559.2 Intron NP_006550.1

View Full Product Details