Product Details

SNP ID
rs116815463
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:110156511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGCAAAAGATTGGAACATACCA[A/G]TATGTCAAAAAGAATATAGCTACAG
Phenotype
MIM: 610299
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC6A17 PubMed Links

Gene Details

Gene
SLC6A17
Gene Name
solute carrier family 6 member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010898.2 Intron NP_001010898.1
XM_006710643.2 Intron XP_006710706.1

View Full Product Details