Product Details

SNP ID
rs117115256
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:84553866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAATTAGGAACTAATAAGGGCA[A/G]CACAATGTTGTTCTTAATTCCCTGG
Phenotype
MIM: 600873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTBS PubMed Links

Gene Details

Gene
CTBS
Gene Name
chitobiase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004388.2 2356 UTR 3 NP_004379.1
Gene
SPATA1
Gene Name
spermatogenesis associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310156.1 2356 Intron NP_001297085.1
XM_011542514.2 2356 Intron XP_011540816.1
XM_011542515.2 2356 Intron XP_011540817.1
XM_011542516.1 2356 Intron XP_011540818.1
XM_011542517.1 2356 Intron XP_011540819.1
XM_011542519.2 2356 Intron XP_011540821.1
XM_011542520.2 2356 Intron XP_011540822.1
XM_011542521.2 2356 Intron XP_011540823.1
XM_011542522.2 2356 Intron XP_011540824.1

View Full Product Details