Product Details

SNP ID
rs117175166
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:168178899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCCGGAAGCAAAAAGCAAAGAG[G/T]CGGAGAAGAAGACGGGAAGCAGAGC
Phenotype
MIM: 611807
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TIPRL PubMed Links

Gene Details

Gene
TIPRL
Gene Name
TOR signaling pathway regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031800.2 55 UTR 5 NP_001026970.1
NM_152902.4 55 UTR 5 NP_690866.1

View Full Product Details