Product Details

SNP ID
rs117303705
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:36341569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAAAAACCAAACAATCGAGCAA[C/T]CATCCCAAAAGGTAGCTTCGTGGTA
Phenotype
MIM: 609437
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STK40 PubMed Links

Gene Details

Gene
STK40
Gene Name
serine/threonine kinase 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282546.1 1944 UTR 3 NP_001269475.1
NM_001282547.1 1944 UTR 3 NP_001269476.1
NM_032017.2 1944 UTR 3 NP_114406.1

View Full Product Details