Product Details

SNP ID
rs117472915
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:40620855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAACAAGTTAGGAAAGATGCTG[C/T]ACATAGAAAATGTATAGAGCACAGA
Phenotype
MIM: 611600
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RIMS3 PubMed Links

Gene Details

Gene
RIMS3
Gene Name
regulating synaptic membrane exocytosis 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014747.2 6696 UTR 3 NP_055562.2
XM_011542479.2 6696 UTR 3 XP_011540781.1

View Full Product Details