Product Details

SNP ID
rs117637379
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:202727950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAAGCTTATTTTAAAAGAGCAC[A/G]GCTTTAAAGATAGGTTCTAGTCCTT
Phenotype
MIM: 605393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDM5B PubMed Links

Gene Details

Gene
KDM5B
Gene Name
lysine demethylase 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314042.1 6994 UTR 3 NP_001300971.1
NM_006618.4 6994 UTR 3 NP_006609.3
XM_011509088.2 6994 UTR 3 XP_011507390.1
XM_011509090.2 6994 UTR 3 XP_011507392.1
XM_011509091.2 6994 UTR 3 XP_011507393.1
XM_011509092.2 6994 UTR 3 XP_011507394.1

View Full Product Details