Product Details

SNP ID
rs117913301
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:247520885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATCTTCTGATTTCCGAAAGGGA[A/G]AAGAAGTTTGTTGCAATCTTCTGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GCSAML PubMed Links

Gene Details

Gene
GCSAML
Gene Name
germinal center associated signaling and motility like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281834.1 Intron NP_001268763.1
NM_001281835.1 Intron NP_001268764.1
NM_001281836.1 Intron NP_001268765.1
NM_001281837.1 Intron NP_001268766.1
NM_001281838.1 Intron NP_001268767.1
NM_001281853.1 Intron NP_001268782.1
NM_145278.4 Intron NP_660321.1
XM_017000399.1 Intron XP_016855888.1
Gene
GCSAML-AS1
Gene Name
GCSAML antisense RNA 1
There are no transcripts associated with this gene.

Gene
OR2C3
Gene Name
olfactory receptor family 2 subfamily C member 3
There are no transcripts associated with this gene.

View Full Product Details