Product Details

SNP ID
rs121909529
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:229431830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACATGACGTTGTTGGCATACAGG[T/A]CCTTCCTGATGTCGATGTCACACTT
Phenotype
MIM: 102610 MIM: 607613
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
ACTA1 PubMed Links

Gene Details

Gene
ACTA1
Gene Name
actin, alpha 1, skeletal muscle
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100.3 986 Missense Mutation GAC,GTC D294V NP_001091.1
Gene
NUP133
Gene Name
nucleoporin 133
There are no transcripts associated with this gene.

View Full Product Details