Product Details

SNP ID
rs121918241
Assay Type
Functionally Tested
NCBI dbSNP Submissions
6
Location
Chr.1:45508329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGGGCTGCTTACTACTACCAA[C/T]GACAAGATGTGGAGGCTGACCCATG
Phenotype
MIM: 609831 MIM: 176763
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCDC163P PubMed Links

Gene Details

Gene
CCDC163P
Gene Name
coiled-coil domain containing 163, pseudogene
There are no transcripts associated with this gene.

Gene
MMACHC
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria
There are no transcripts associated with this gene.

Gene
PRDX1
Gene Name
peroxiredoxin 1
There are no transcripts associated with this gene.

View Full Product Details