Product Details
- SNP ID
-
rs137853967
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:16986864 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGGACACGGCTGCAGCCAGGATG[C/A]GGTACTGGAAGCTGGACAGAGAGAA
- Phenotype
-
MIM: 610513
MIM: 156790
- Polymorphism
- C/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ATP13A2
PubMed Links
Gene Details
- Gene
- ATP13A2
- Gene Name
- ATPase 13A2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001141973.2 |
3383 |
Missense Mutation |
CGC,CTC |
R1054L |
NP_001135445.1 |
NM_001141974.2 |
3383 |
Missense Mutation |
CGC,CTC |
R1015L |
NP_001135446.1 |
NM_022089.3 |
3383 |
Missense Mutation |
CGC,CTC |
R1059L |
NP_071372.1 |
XM_005245810.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1058L |
XP_005245867.1 |
XM_005245811.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1054L |
XP_005245868.1 |
XM_005245812.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1050L |
XP_005245869.1 |
XM_005245815.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1020L |
XP_005245872.1 |
XM_006710512.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1053L |
XP_006710575.1 |
XM_006710513.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1045L |
XP_006710576.1 |
XM_011541128.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1054L |
XP_011539430.1 |
XM_011541129.1 |
3383 |
Missense Mutation |
CGC,CTC |
R990L |
XP_011539431.1 |
XM_017000844.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1054L |
XP_016856333.1 |
XM_017000845.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1053L |
XP_016856334.1 |
XM_017000846.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1045L |
XP_016856335.1 |
XM_017000847.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1044L |
XP_016856336.1 |
XM_017000848.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1020L |
XP_016856337.1 |
XM_017000849.1 |
3383 |
Missense Mutation |
CGC,CTC |
R1015L |
XP_016856338.1 |
XM_017000850.1 |
3383 |
Missense Mutation |
CGC,CTC |
R990L |
XP_016856339.1 |
- Gene
- MFAP2
- Gene Name
- microfibrillar associated protein 2
There are no transcripts associated with this gene.
View Full Product Details