Product Details

SNP ID
rs137856818
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:27393966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCTCAGGCACCCTGGTGTCCTG[C/T]GAGAATGCGCTAGTGGTGGCCATCA
Phenotype
MIM: 600241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR3 PubMed Links

Gene Details

Gene
GPR3
Gene Name
G protein-coupled receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005281.3 267 Silent Mutation TGC,TGT C56C NP_005272.1

View Full Product Details