Product Details

SNP ID
rs137902829
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:206899404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGGTTTTACCCTGCTTCTCTG[A/G]AGCCAGGTATCAGGGGCCCAGGGCC
Phenotype
MIM: 606015 MIM: 604136
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FCMR PubMed Links

Gene Details

Gene
FCMR
Gene Name
Fc fragment of IgM receptor
There are no transcripts associated with this gene.

Gene
IL24
Gene Name
interleukin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185156.1 404 Nonsense Mutation TGA,TGG *44W NP_001172085.1
NM_001185157.1 404 Nonsense Mutation TGA,TGG *44W NP_001172086.1
NM_001185158.1 404 Intron NP_001172087.1
NM_006850.3 404 Nonsense Mutation TGA,TGG *43W NP_006841.1
XM_011509101.2 404 Nonsense Mutation TGA,TGG *16W XP_011507403.1
XM_017000121.1 404 Nonsense Mutation TGA,TGG *16W XP_016855610.1

View Full Product Details