Product Details

SNP ID
rs137927189
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:162074690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCTTATGAATCATGGCTCAGGA[A/G]TTCAGGCAGGGCTTGGCTGGGCTGT
Phenotype
MIM: 605551
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOS1AP PubMed Links

Gene Details

Gene
NOS1AP
Gene Name
nitric oxide synthase 1 adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126060.1 Intron NP_001119532.2
NM_001164757.1 Intron NP_001158229.1
NM_014697.2 Intron NP_055512.1

View Full Product Details