Product Details

SNP ID
rs137952140
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:52687795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGCTCGATTTTTTAGCACCTC[A/G]GCCTTGGCCTCATCCTTATCAACAC
Phenotype
MIM: 615623
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COA7 PubMed Links

Gene Details

Gene
COA7
Gene Name
cytochrome c oxidase assembly factor 7 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023077.2 661 Silent Mutation GCC,GCT A207A NP_075565.2

View Full Product Details