Product Details

SNP ID
rs138021584
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:169515483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATTTCAGCCTGTATGGTTTCCA[C/T]TCCACTCCCTGCTCACTGTAGTGGA
Phenotype
MIM: 612309
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F5 PubMed Links

Gene Details

Gene
F5
Gene Name
coagulation factor V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000130.4 6634 Silent Mutation GAA,GAG E2163E NP_000121.2
XM_017000660.1 6634 Silent Mutation GAA,GAG E2026E XP_016856149.1

View Full Product Details