Product Details

SNP ID
rs138028716
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:153960631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCACCATTCACTGTTCCCAGCA[A/G]AGCCCTTGTTTCCTCCAGAGGTGAC
Phenotype
MIM: 607138 MIM: 608972 MIM: 604740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CREB3L4 PubMed Links

Gene Details

Gene
CREB3L4
Gene Name
cAMP responsive element binding protein 3 like 4
There are no transcripts associated with this gene.

Gene
CRTC2
Gene Name
CREB regulated transcription coactivator 2
There are no transcripts associated with this gene.

Gene
MIR6737
Gene Name
microRNA 6737
There are no transcripts associated with this gene.

Gene
SLC39A1
Gene Name
solute carrier family 39 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271957.1 790 Silent Mutation CTG,TTG L148L NP_001258886.1
NM_001271958.1 790 Silent Mutation CTG,TTG L148L NP_001258887.1
NM_001271959.1 790 Silent Mutation CTG,TTG L148L NP_001258888.1
NM_001271960.1 790 Silent Mutation CTG,TTG L148L NP_001258889.1
NM_001271961.1 790 Missense Mutation TCT,TTT S104F NP_001258890.1
NM_014437.4 790 Silent Mutation CTG,TTG L148L NP_055252.2

View Full Product Details