Product Details
- SNP ID
-
rs138045281
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:156468202 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGGAGGGGGAGGCGCAGGGCTGC[A/G]CTCACGGCTTGGGGACACCGGTTCT
- Phenotype
-
MIM: 600663
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MEF2D
PubMed Links
Gene Details
- Gene
- MEF2D
- Gene Name
- myocyte enhancer factor 2D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001271629.1 |
1812 |
Missense Mutation |
CGC,TGC |
R442C |
NP_001258558.1 |
NM_005920.3 |
1812 |
Missense Mutation |
CGC,TGC |
R449C |
NP_005911.1 |
XM_005245169.4 |
1812 |
Missense Mutation |
CGC,TGC |
R449C |
XP_005245226.1 |
XM_005245170.3 |
1812 |
Missense Mutation |
CGC,TGC |
R449C |
XP_005245227.1 |
XM_006711330.3 |
1812 |
Missense Mutation |
CGC,TGC |
R449C |
XP_006711393.1 |
XM_006711332.3 |
1812 |
Missense Mutation |
CGC,TGC |
R448C |
XP_006711395.1 |
XM_006711333.2 |
1812 |
Missense Mutation |
CGC,TGC |
R442C |
XP_006711396.1 |
XM_006711334.3 |
1812 |
Missense Mutation |
CGC,TGC |
R441C |
XP_006711397.1 |
XM_011509569.2 |
1812 |
Missense Mutation |
CGC,TGC |
R449C |
XP_011507871.1 |
XM_017001314.1 |
1812 |
Missense Mutation |
CGC,TGC |
R442C |
XP_016856803.1 |
XM_017001315.1 |
1812 |
Missense Mutation |
CGC,TGC |
R442C |
XP_016856804.1 |
View Full Product Details