Product Details

SNP ID
rs138052296
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:17069220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCCAGGCAGCATTCCTCCTCAA[C/T]CTGTGGCCCGAATGGCTTGGGGATG
Phenotype
MIM: 607935
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PADI2 PubMed Links

Gene Details

Gene
PADI2
Gene Name
peptidyl arginine deiminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007365.2 1078 Missense Mutation ATT,GTT I608V NP_031391.2
XM_017000148.1 1078 Missense Mutation ATT,GTT I293V XP_016855637.1

View Full Product Details