Product Details
- SNP ID
-
rs138105602
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:152213168 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAAATATGAGCCAGAACTTCCCC[C/T]ATCATGGTTACTTCCTCCTTTGCAA
- Phenotype
-
MIM: 616293
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HRNR
PubMed Links
Gene Details
- Gene
- HRNR
- Gene Name
- hornerin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001009931.2 |
8540 |
Missense Mutation |
AGG,GGG |
R2821G |
NP_001009931.1 |
View Full Product Details