Product Details

SNP ID
rs138129705
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:158547454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGCAGCGTAGGATGCCACCACAA[C/T]ACAAAGAGGAATAGCAATGACCATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6Y1 PubMed Links

Gene Details

Gene
OR6Y1
Gene Name
olfactory receptor family 6 subfamily Y member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005189.1 652 Missense Mutation ATT,GTT I218V NP_001005189.1

View Full Product Details